Learn More
Aviva Systems Biology Corp ATN1 Antibody - N-terminal region (ARP34153_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34153_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATN1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ATN1. Alternate names: B37, D12S755E, DRPLA, NOD, HRSApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55273