Learn More
Medchemexpress LLC Atomoxetine (hydrochloride) | 82248-59-7 | 99.9% | 291.82 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1738510MM/1ML
Encompass_Preferred
Atomoxetine hydrochloride (Tomoxetine hydrochloride) is a selective noradrenaline reuptake inhibitor with Ki values of 5 nM, 77 nM, and 1451 nM for norepinephrine (NE), serotonin (5-HT), and dopamine (DA) transporters, respectively. It is also a potent Na+ channels (VGSCs) blocker and can be used for attention-deficit hyperactivity disorder (ADHD) research.
  • Selective noradrenaline reuptake inhibitor
  • Potent Na+ channels (VGSCs) blocker
  • Used for attention-deficit hyperactivity disorder (ADHD) research
  • High purity: 99.91%
Catalog No. 50-003-84241