Learn More
Medchemexpress LLC Atomoxetine hydrochloride | 82248-59-7 | 99.91% | 291.82 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY17385
Encompass_Preferred
Atomoxetine hydrochloride, also known as Tomoxetine hydrochloride, is a selective noradrenaline reuptake inhibitor. It demonstrates Ki values of 5 nM, 77 nM, and 1451 nM for norepinephrine (NE), serotonin (5-HT), and dopamine (DA) transporters, respectively. This compound also acts as a potent Na+ channels (VGSCs) blocker.
  • Selective noradrenaline reuptake inhibitor.
  • Potent Na+ channels (VGSCs) blocker.
  • Utilized for research related to attention-deficit hyperactivity disorder (ADHD).
Catalog No. 50-003-84325