Learn More
Medchemexpress LLC Atomoxetine hydrochloride | 82248-59-7 | MFCD06410992 | 99.9% | 291.82 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1738550MG
Encompass_Preferred
Atomoxetine hydrochloride is a selective noradrenaline reuptake inhibitor with Ki values of 5 nM, 77 nM and 1451 nM for norepinephrine (NE), serotonin (5-HT) and dopamine (DA) transporters, respectively. It is also a potent Na+ channels (VGSCs) blocker and can be used for attention-deficit hyperactivity disorder (ADHD) research.
  • Selective noradrenaline reuptake inhibitor.
  • Potent Na+ channels (VGSCs) blocker.
  • Can be used for attention-deficit hyperactivity disorder (ADHD) research.
  • Available in solid and solution forms.
  • Purity: 99.91%.
Catalog No. 50-003-85220