Learn More
TARGETMOL CHEMICALS INC ATOPAXAR 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T19865MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Atopaxar (E5555) a reversible PAR-1 thrombin receptor antagonist interferes with platelet signaling. Atopaxar has been investigated for the treatment of Coronary Artery Disease and Acute Coronary Syndrome. purity: 98%
Catalog No. 50-257-7254