Learn More
Selleck Chemical LLC Atorvastatin Calcium 50mg 134523-03-8
SDP
Supplier:  Selleck Chemical LLC S207750MG
Encompass_Preferred
Atorvastatin Calcium is an inhibitor of HMG-CoA reductase used as a cholesterol-lowering medication that blocks the production of cholesterol. Atorvastatin Calcium induces apoptosis and autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0980320