Learn More
Medchemexpress LLC Atovaquone | 95233-18-4 | 366.84 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1383210MM/1ML
Encompass_Preferred
Atovaquone is a potent, selective, and orally active inhibitor of the parasite's mitochondrial cytochrome bc1 complex. It is effective against human and P. falciparum cytochrome bc1 activity. Atovaquone is an antimalarial agent with potential for investigating pneumocystis pneumonia, toxoplasmosis, malaria, and babesia.
  • Potent, selective, and orally active inhibitor
  • Targets the Qo site of the Plasmodium cytochrome bc1 complex
  • Demonstrates an IC50 of 67 nM for transmission blocking potential
  • Improved mice survival rate in animal models
Catalog No. 50-003-50264