Learn More
Medchemexpress LLC Atovaquone | 95233-18-4 | 99.8% | 366.84 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY13832100MG
Encompass_Preferred
Atovaquone is a potent, selective, and orally active inhibitor of the parasite's mitochondrial cytochrome bc1 complex. It is an antimalarial agent with potential for investigating pneumocystis pneumonia, toxoplasmosis, malaria, and babesia.
  • Potent, selective, and orally active inhibitor of the parasite's mitochondrial cytochrome bc1 complex.
  • Effective against human and P. falciparum cytochrome bc1 activity.
  • Active against development in the mosquito from gamete production to ookinete.
  • Potential for investigating pneumocystis pneumonia, toxoplasmosis, malaria, and babesia.
Catalog No. 50-003-50459