Learn More
Eurofins Discoverx ATP, 10 mg. (92-1176)
Supplier:  Eurofins Discoverx 921176
Encompass
Adenosine 5′-triphosphate (ATP) is a multifunctional nucleotide and a key component of energy storage and metabolism used in many cellular processes. Serves as a substrate for kinases and coenzymes for a wide array of enzymatic reactions in-vivo. Potent p2 purinoceptor agonist. Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9704