Learn More
Apexbio Technology LLC ATP(Synonyms: Adenosine triphosphate, Adenosine 5'-triphosphate, Adenosine-5'-triphosphoric acid), 500mg, CAS: 56-65-5.
SDP
Supplier:  Apexbio Technology LLC C6931500
Encompass_Preferred
Adenosine triphosphate (ATP CAS 56-65-5) is a nucleoside triphosphate comprising adenine linked to ribose with three phosphate groups esterified sequentially ATP primarily functions as a universal energy carrier in cellular metabolism transferring phosphate groups to facilitate enzymatic reactions and drive biological activities Additionally ATP acts extracellularly as a neurotransmitter and signaling molecule binding purinergic receptors to modulate physiological responses such as neurotransmission vascular tone inflammation and immune cell function Due to its versatile biological roles ATP is extensively employed in biomedical research investigating metabolic pathways receptor signaling dynamics and cellular energetics
Catalog No. 50-000-61171