Learn More
Medchemexpress LLC ATP dimagnesium | 74804-12-9 | 98.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB2176B100MG
Encompass_Preferred
ATP (Adenosine 5'-triphosphate) dimagnesium is a vital element in energy storage and metabolism within living organisms. It supplies the metabolic energy required for metabolic pumps and functions as a coenzyme in cells. Additionally, ATP dimagnesium acts as a significant endogenous signaling molecule involved in immunity and inflammation.
  • A central component of energy storage and metabolism in vivo.
  • Provides metabolic energy to drive metabolic pumps.
  • Serves as a coenzyme in cells.
  • An important endogenous signaling molecule in immunity and inflammation.
  • For research use only.
Catalog No. 50-004-10470