Learn More
Selleck Chemical LLC ATP disodium trihydrate E4964-1G
SDP
Supplier:  Selleck Chemical LLC E49641G
Encompass_Preferred
ATP disodium trihydrate is the primary energy currency of the cell essential for intracellular energy metabolism It regulates extracellular biological processes including cardiac function neurotransmission muscle contraction vasodilation bone and liver glycogen metabolism and inflammation It also plays a crucial role in initiating immune responses during infections
Catalog No. 50-000-42124