Learn More
Abcam ATP Stock Solution, 200UL
Supplier:  Abcam AB189136200UL
Encompass_Preferred
10 mM ATP stock solution used for preparing hot ATP assay cocktail.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4398