Learn More
Medchemexpress LLC ATP synthase inhibitor 1 | 1023043-30-2 | 99.56% | 411.93 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11271510MG
Encompass_Preferred
ATP synthase inhibitor 1 is a potent inhibitor of the c subunit of the F1/FO-ATP synthase complex. It inhibits mitochondrial permeability transition pore (mPTP) opening and does not affect ATP levels. It is intended for research use only.
  • Potent inhibitor of the c subunit of the F1/FO-ATP synthase complex
  • Inhibits mitochondrial permeability transition pore (mPTP) opening
  • Does not affect ATP levels
Catalog No. 50-004-38532