Learn More
Medchemexpress LLC ATP synthase inhibitor 1 | 1023043-30-2 | 99.6% | 411.93 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY112715100MG
Encompass_Preferred
ATP synthase inhibitor 1 is a potent inhibitor of the c subunit of the F1/FO-ATP synthase complex. It inhibits mitochondrial permeability transition pore (mPTP) opening and does not affect ATP levels. This product is for research use only.
  • Potent inhibitor of the c subunit of the F1/FO-ATP synthase complex.
  • Inhibits mitochondrial permeability transition pore (mPTP) opening.
  • Does not affect ATP levels.
  • For research use only.
Catalog No. 50-004-38551