Learn More
PCE AMERICAS INC IoT Sensor Stores up to 10,000 date- and time-stamped data records to internal memory
SDP
Supplier:  PCE AMERICAS INC PCEATP 1
Encompass_Preferred
The PCE-ATP 1 is used to quickly and easily check the cleanliness of surfaces. The Hygiene Tester displays a measurement result on the large LC display after only 10 seconds. The Hygiene Tester's internal memory stores up to 10,000 measured data sets with time and date. The PCE-ATP 1 with the associated PCE-ATP SwapPens, allows a quick detection of food or microbiological residues on all surfaces in contact with the product as an effectiveness test of cleaning and disinfection measures. Quantitative determination of intracellular and extracellular adenosine triphosphate (ATP) is performed in the portable PCE-ATP 1. The measured data from the Hygiene Tester can be stored and further processed after the measurement. The Hygiene Tester is used in the hygiene control in pharmacy, medicine, gastronomy, wholesalers and more.
Catalog No. NC3654615