Learn More
Biomatik Corporation Atp8B1 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC10929
Encompass_Preferred
Atp8B1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to ATP8B1. The Atp8B1 Polyclonal Antibody has been validated for the following applications: ELISA, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Phospholipid-Transporting Atpase Ic Protein (217-330Aa). Applications: ELISA, IF. Target Synonyms: ATP8B1. Accession Number: O43520. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9255