Learn More
TARGETMOL CHEMICALS INC Atpenin A5 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T97141MG
Encompass_Preferred
Also available in 2 mg, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Atpenin A5 is a potent and highly specific complex II inhibitor (IC50 ~10 nM), as well as an effective mKATP channel agonist and cardioprotective agent [1]. Purity 95%
Catalog No. 50-287-7233