Learn More
FOCUS BIOMOLECULES AtPep1, Innate immune response activator, >95% by HPLC
SDP
Catalog No. NC3199235
Encompass
Biochemical Reagents, AtPep1, Innate immune response activator, >95% by HPLC, Sequence: ATKVKAKQRGKEKVSSGRPGQHN; AtPep1 is an endogenous peptide elicitor from Arabidopsis leaves that signals the activation of components of the innate immune response against pathogens. Atpep1 derives from the carboxyl terminal of the precursor propeptide PROPEP1, and on microbe-associated molecular pattern (MAMP) perception or cell damage, vacuolar membrane-resident PROPEP1 is processed by type II metacaspases to release AtPep1 which binds to the leucine-rich repeat receptor kinase (LLR-RK) PEPR1. AtPep1 binding to PEPR1 leads to heterodimerization with the shared receptor kinase Brassinosteroid-Insensitive 1-Associated Receptor Kinase 1 (BAK1) and consequent activation of the receptor complex, resulting in immune responses similar to those induced by pathogens.
Catalog No. NC3199235 Supplier FOCUS BIOMOLECULES Supplier No. PL030
Biochemical Reagents, AtPep1, Innate immune response activator, >95% by HPLC, Sequence: ATKVKAKQRGKEKVSSGRPGQHN; AtPep1 is an endogenous peptide elicitor from Arabidopsis leaves that signals the activation of components of the innate immune response against pathogens. Atpep1 derives from the carboxyl terminal of the precursor propeptide PROPEP1, and on microbe-associated molecular pattern (MAMP) perception or cell damage, vacuolar membrane-resident PROPEP1 is processed by type II metacaspases to release AtPep1 which binds to the leucine-rich repeat receptor kinase (LLR-RK) PEPR1. AtPep1 binding to PEPR1 leads to heterodimerization with the shared receptor kinase Brassinosteroid-Insensitive 1-Associated Receptor Kinase 1 (BAK1) and consequent activation of the receptor complex, resulting in immune responses similar to those induced by pathogens.