Learn More
Medchemexpress LLC HY-136270 5mg Medchemexpress, ATR inhibitor 2 CAS:1613191-99-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1362705MG
Encompass_Preferred
Medchemexpress, HY-136270 5mg ATR inhibitor 2 CAS:1613191-99-3 ATR inhibitor 2 is an ATP-competitive, orally active, and selective ATR inhibitor, with a Ki of <150 pM. ATR inhibitor 2 potently inhibits ATR-driven phosphorylated checkpoint kinase-1 (Chk1) phosphorylation with an IC50 of 8 nM. Antitumor activity[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0413