Learn More
Medchemexpress LLC ATRA-biotin | 2226143-93-5 | >98.43% | C36H55N3O4S | 50 MG
Supplier: Medchemexpress LLC HY14179350MG
ATRA-biotin, also known as Biotin-ATRA-conjugate, is a biotin-conjugated derivative of All-Trans Retinoic Acid (ATRA). This compound serves as a valuable tool for tracking ATRA within cellular environments or specific tissues, enabling researchers to investigate its distribution and interactions.
- Biotin-conjugated derivative of All-Trans Retinoic Acid (ATRA)
- Used to track ATRA in cells or a given tissue
- Targets RAR/RXR, PPAR, Endogenous Metabolite, and Autophagy pathways
- Soluble in DMSO at 100 mg/mL
- Demonstrates biocompatibility in cell viability assays