Learn More
Cayman Chemical AtractylenolIde II 5mg
SDP
Supplier:  Cayman Chemical 361735 MG
Encompass_Preferred
A sesquiterpene with diverse biological activities; inhibits proliferation of, as well as halts the cell cycle at the G2/M phase and induces apoptosis in, DU145 and LNCaP prostate cancer cells at 50 and 100 µM; prevents irradiation-induced cutaneous and intestinal ulcers in mice at 60 mg/kg
Catalog No. 50-291-2004