Learn More
TARGETMOL CHEMICALS INC ATRACTYLODIN 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T366610MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Atractylodin (Atractydin) has high lipase inhibitory activity (IC539.12 uM); Atractylodin possesses strong repellency activities against T. castaneum adults (LD50 1.83 ug/cm). The repellency of Atractylodin (0.63 ug/cm) can achieve >90%. purity: 99%
Catalog No. 50-257-6890