Learn More
Medchemexpress LLC Atractyloside potassium salt | 102130-43-8 | 802.99 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN146210MM/1ML
Encompass_Preferred
Atractyloside potassium salt is a powerful and specific inhibitor of mitochondrial ADP/ATP transport. It inhibits chloride channels from mitochondrial membranes of rat heart, activates autophagy, inhibits ANT2, mTOR, and promotes the activation of p-AMPK. This compound exhibits anti-cancer effects on non-small cell lung cancer and can inhibit liver steatosis. However, it also has nephrotoxicity.
  • Powerful and specific inhibitor of mitochondrial ADP/ATP transport.
  • Inhibits chloride channels from mitochondrial membranes of rat heart.
  • Activates autophagy, inhibits ANT2, mTOR, and promotes the activation of p-AMPK.
  • Exhibits anti-cancer effects on non-small cell lung cancer.
  • Can inhibit liver steatosis.
  • Purity: 99.93%.
Catalog No. 50-004-14645