Learn More
Cayman Chemical AtractylosIdepotasIM salt 10mg
SDP
Supplier:  Cayman Chemical 1480410 MG
Encompass_Preferred
A natural heteroglucoside that blocks mitochondrial ADP/ATP translocases, inhibiting oxidative phosphorylation
Catalog No. 50-297-1955