Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Atrazine-d5 | 163165-75-1 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYN7091S25MG

Encompass_Preferred

Atrazine-d5 is deuterium labeled Atrazine. Atrazine is primarily used for controlling certain annual broadleaf and grass weeds. It inhibits photophosphorylation but typically does not cause lethality or permanent cell damage in the short term.

  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Molecular weight: 220.71
  • Formula: C8H9D5ClN5
  • Appearance: Solid
  • Color: White to off-white
  • Purity: 98.72%

Catalog No. 50-004-18702


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.