Learn More
Cayman Chemical AtrIal NatrIuretIc PeptID3 1mg
SDP
Supplier:  Cayman Chemical 245401 MG
Encompass_Preferred
An atrial natriuretic peptide; a synthetic and truncated form of ANP; potently inhibits renin release from cultured renal juxtaglomerular cells (Ki = 10 pM) in a cGMP-dependent manner; dose-dependently decreases MAP and arterial plasma renin activity and increases fractional sodium and calcium excretion, urine volume, and glomerular filtration rate in conscious dogs (maximum effect = 200 pM)
Catalog No. 50-293-4917