Learn More
CPC Scientific H-Arg-Arg-Ser-Ser-Cys-Phe-Gly-Gly-Arg-Met-Asp-Arg-Ile-Gly-Ala-Gln-Ser-Gly-Leu-Gly-Cys-Asn-Ser-Phe-Arg-Tyr-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific NATR002B
Encompass
ONE-LETTER SEQUENCE: RRSSCFGGRMDRIGAQSGLGCNSFRY (Cys5 and 21 bridge)MOLECULAR FORMULA: C118H187N43O36S3MOLECULAR WEIGHT:2880.25STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [102686-43-1], SYNONYMS: hANF (3-28), Atrial Natriuretic Factor (3-28) (human, bovine, porcine)RESEARCH AREA: Cardiovascular
Catalog No. 50-210-9850