Learn More
Medchemexpress LLC Atrial natriuretic peptide (3-28) (human) | 102686-43-1 | 99.7% | 2880.21 | 5 MG

Supplier: Medchemexpress LLC HYP57925MG
Atrial natriuretic peptide (3-28) (human) is a peptide hormone synthesized and secreted by the atrial myocardium. It is involved in the regulation of blood pressure, fluid balance, and electrolyte homeostasis.
- High purity of 99.65%
- Molecular weight of 2880.21
- Chemical formula C118H187N43O36S3
- Appears as a white to off-white solid
- Specific amino acid sequence: Arg-Arg-Ser-Ser-Cys-Phe-Gly-Gly-Arg-Met-Asp-Arg-Ile-Gly-Ala-Gln-Ser-Gly-Leu-Gly-Cys-Asn-Ser-Phe-Arg-Tyr (disulfide bridge: Cys5-Cys21)
- Regulates blood pressure, fluid balance, and electrolyte homeostasis
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.