Learn More
CPC Scientific H-Ser-Ser-Cys-Phe-Gly-Gly-Arg-Ile-Asp-Arg-Ile-Gly-Ala-Gln-Ser-Gly-Leu-Gly-Cys-Asn-Ser-Phe-Arg-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific ATPE002B
Encompass
Atriopeptin II elevates cyclic GMP, activates cyclic GMP- dependent protein kinase and Atriopeptin II causes relaxation in rat thoracic aorta.ONE-LETTER SEQUENCE: SSCFGGRIDRIGAQSGLGCNSFR(Cys&Cys bridge)MOLECULAR FORMULA: C98H156N34O32S2MOLECULAR WEIGHT:2386.66STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [89139-54-8], RESEARCH AREA: CardiovascularREFERENCES: M.G.Currie et al., Science, 223, 67 (1984)
Catalog No. 50-210-8831