Learn More
CPC Scientific H-Ser-Ser-Cys-Phe-Gly-Gly-Arg-Ile-Asp-Arg-Ile-Gly-Ala-Gln-Ser-Gly-Leu-Gly-Cys-Asn-Ser-Phe-Arg-Tyr-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific ATPE003A
Encompass
Atriopeptin III inhibits carotid body chemoreceptor nerve activity induced by hypoxia.ONE-LETTER SEQUENCE: SSCFGGRIDRIGAQSGLGCNSFRY(Cys&Cys bridge)MOLECULAR FORMULA: C107H165N35O34S2MOLECULAR WEIGHT:2549.83STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [90817-13-3], RESEARCH AREA: CardiovascularREFERENCES: D.M.Geller et al., Biochem. Biophys. Res. Commun., 120, 333 (1984)
Catalog No. 50-210-8832