Learn More
Medchemexpress LLC Atropine | 51-55-8 | 99.85% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB1205500MG
Encompass_Preferred
Atropine is a competitive antagonist of muscarinic acetylcholine receptors (mAChR), demonstrating IC50 values of 0.39 and 0.71 nM for human mAChR M4 and chicken mAChR M4, respectively. It works by inhibiting acetylcholine-induced relaxations in human pulmonary veins.
  • Acts as a competitive muscarinic acetylcholine receptor antagonist.
  • Inhibits acetylcholine-induced relaxations in human pulmonary veins.
  • Can be used for research related to anti-myopia.
  • Can be used for research related to bradycardia.
Catalog No. 50-004-12233