Learn More
Medchemexpress LLC Atropine | 51-55-8 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB120510MM/1ML
Encompass_Preferred
Atropine (Tropine tropate) is a competitive muscarinic acetylcholine receptor (mAChR) antagonist. It inhibits ACh-induced relaxations in human pulmonary veins and can be used for research on anti-myopia and bradycardia.
  • Competitive muscarinic acetylcholine receptor (mAChR) antagonist.
  • IC50 values of 0.39 and 0.71 nM for human mAChR M4 and chicken mAChR M4.
  • Inhibits ACh-induced relaxations in human pulmonary veins.
  • Used for research of anti-myopia and bradycardia.
Catalog No. 50-004-11146