Learn More
Medchemexpress LLC Atropine oxide hydrochloride | 4574-60-1 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY122510A100MG
Encompass_Preferred
Atropine oxide hydrochloride is the hydrochloride salt of atropine N-oxide, a muscarinic acetylcholine receptor antagonist used in pharmacology and toxicology research. It is commonly used to study muscarinic receptor activity and in research on treatments for nerve agent or pesticide exposure.
  • Acts as a competitive antagonist at M1-M5 muscarinic receptors.
  • Supplied as the hydrochloride salt for improved stability and handling.
  • Available in small milligram quantities for research use.
  • Suitable for receptor pharmacology and toxicology studies.
Catalog No. 50-002-32586