Learn More
Selleck Chemical LLC Atropine sulfate monohydrate 10mg 5908-99-6
SDP
Supplier:  Selleck Chemical LLC S213010MG
Encompass_Preferred
Atropine sulfate monohydrate is a competitive antagonist for the muscarinic acetylcholine receptor, used to decrease the production of saliva and secretions of the airway prior to surgery. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-541-6