Learn More
MSC Nilfisk Wet/Dry Vacuum, 30 L, Electric, 7.4 Amp
Supplier:  MSC 38686325
Encompass
The ATTIX 33 wet/dry vacuum is designed for professionals, offering high performance, increased productivity, and lower maintenance. It has a 30 liter capacity and auto tool start.
  • InfiniClean system automatically cleans the filter during operation, maintaining high performance.
  • Automatic tool start activates dust extraction, continuing briefly after power-off for total dust containment.
  • Durable, washable PTFE filter ensures minimum 99.9% filtration of fine dust particles.
  • Flat top provides storage for safety gear, accessories, or power tools.
  • Double-walled construction ensures long product life in tough environments.
  • Easy-to-use hooks and flexible rubber straps for simple storage and transportation.
  • Large wheels, locking swivel casters, and optional handle provide easy maneuverability.
  • Optional universal toolbox mount available.
Catalog No. 50-283-6059