Learn More
Abcam Atto 488 Conjugation Kit (Fast) - Lightning-Link, 3 x 100UG
Supplier:  Abcam AB2698963X100UG
Encompass_Preferred
ATTO 488 conjugation / labeling in < 20 mins with 30 secs hands-on time using Atto 488 Conjugation Kit (Fast) - Lightning-Link ab269896. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2022527