Learn More
Medchemexpress LLC Atuveciclib | 2923012-24-0 | 99.8% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY12871B100MG
Encompass_Preferred
Atuveciclib is a potent and highly selective oral PTEFb/CDK9 inhibitor that targets CDK9/CycT1 with an IC50 of 13 nM. It demonstrates potent antiproliferative activity against various cell lines, including HeLa (IC50=920 nM) and MOLM-13 (IC50=310 nM) in vitro. In vivo studies in the MOLM-13 xenograft model show high antitumor efficacy with dose-dependent results and good tolerability, with less than 10% mean body weight reduction. It also exhibits low blood clearance in rats.
  • Potent and highly selective oral PTEFb/CDK9 inhibitor
  • Inhibits CDK9/CycT1 with an IC50 of 13 nM
  • Demonstrates potent antiproliferative activity against various cell lines
  • Shows high antitumor efficacy in MOLM-13 xenograft model
  • Well-tolerated in vivo studies
  • Exhibits low blood clearance in rats
Catalog No. 50-003-42537