Learn More
Selleck Chemical LLC Atuveciclib S8727-10MM/1ML
SDP
Supplier:  Selleck Chemical LLC S872710MM/1ML
Encompass_Preferred
Atuveciclib (BAY-1143572) is a potent and highly selective PTEFb/CDK9 inhibitor with IC50 values of 13 nM for CDK9/CycT and the ratio of IC50 values for CDK2/CDK9 is about 100 Outside the CDK family It inhibits GSK3 kinase with IC50 values of 45 nM and 87 nM for GSK3 and GSK3 respectively
Catalog No. 50-000-24433