Learn More
Selleck Chemical LLC Atuveciclib (BAY-1143572) 2mg 1414943-88-6
SDP
Supplier:  Selleck Chemical LLC S87272MG
Encompass_Preferred
Atuveciclib (BAY-1143572) is potent and highly selective PTEFb/CDK9 inhibitor with IC50 values of 13 nM for CDK9/CycT and the ratio of IC50 values for CDK2/CDK9 is about 100. Outside the CDK family, It inhibits GSK3 kinase with IC50 values of 45 nM and 87 nM for GSK3? and GSK3? respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8169