Learn More
Medchemexpress LLC ATX inhibitor 1 | 2225892-70-4 | 99.3% | 501.30 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY11141050MG
Encompass_Preferred
ATX inhibitor 1 is a potent autotaxin inhibitor with IC50 values of 1.23 nM (FS-3) and 2.18 nM (bis-pNPP).
  • Potent inhibitor of autotaxin.
  • For research use only.
  • Available in solid and solution forms.
  • High purity.
  • Detailed solubility and storage information provided.
Catalog No. 50-003-44024