Learn More
Medchemexpress LLC ATX inhibitor 1 | 2225892-70-4 | 99.3% | 501.30 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY11141010MM/1ML
Encompass_Preferred
ATX inhibitor 1 is a potent ATX inhibitor designed for research applications. It effectively inhibits ATX with IC50 values of 1.23 nM (FS-3) and 2.18 nM (bis-pNPP). This product is intended for laboratory use only.
  • Potent ATX inhibitor
  • Suitable for research applications
  • Available in solid and solution formulations
  • Soluble in DMSO for in vitro studies
  • Stable for long-term storage when handled correctly
Catalog No. 50-003-43564