Learn More
Enzo Life Sciences AU1 monoclonal antibody (AU1) (purified) (200µl)
SDP
Supplier:  Enzo Life Sciences ENZABS1350200
Encompass_Preferred
Clone: AU1. Isotype: IgG2a. Immunogen: Raised against an SDS-disrupted BPV-1 preparation and mapped to the six amino acid epitope DTYRYI. Specificity: Recognizes the the six amino acid epitope DTYRYI. Applications: IF, IP, WB. Purity: Purified. Formulation: Liquid. In PBS containing 0.03% thimerosal.
Catalog No. 50-201-2698