Learn More
TARGETMOL CHEMICALS INC AUCUPARIN 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TN34695MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Aucuparin (26-dimethoxy-4-phenylphenol) is a plant antibiotic with anti-inflammatory activity. Aucuparin inhibited pulmonary fibrosis in mouse models induced by bleomycin (BLM). ABTS and FRAP showed clear scavenging activity. Aucuparin inhibited superoxide production of human neutrophils induced by fmlp and its IC50 was 17.0uM. purity: 99%
Catalog No. 50-257-8874