Learn More
Medchemexpress LLC AUNP-12 (NP-12) | 1353563-85-5 | 99.2% | 3261.55 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP18121MG
Encompass_Preferred
AUNP-12 (NP-12) is a peptide antagonist of the PD-1 signaling pathway, demonstrating equipotent antagonism toward PD-L1 and PD-L2 in the rescue of lymphocyte proliferation and effector functions. It exhibits immune activation, excellent antitumor activity, and potential for better management of immune-related adverse events (irAEs).
  • Peptide antagonist of PD-1 signaling pathway
  • Equipotent antagonism toward PD-L1 and PD-L2
  • Rescues lymphocyte proliferation and effector functions
  • Exhibits immune activation and antitumor activity
  • Potential for better management of immune-related adverse events
Catalog No. 50-004-24088