Learn More
Medchemexpress LLC AUNP-12 (NP-12) | 1353563-85-5 | 99.2% | 3261.55 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP18125MG
Encompass_Preferred
AUNP-12 is a peptide antagonist of the PD-1 signaling pathway. It demonstrates equipotent antagonism towards PD-L1 and PD-L2, rescuing lymphocyte proliferation and effector functions. This peptide shows immune activation and significant antitumor activity, offering potential for improved management of immune-related adverse events (irAEs). It is intended for research use only.
  • Peptide antagonist of the PD-1 signaling pathway
  • Demonstrates equipotent antagonism towards PD-L1 and PD-L2
  • Rescues lymphocyte proliferation and effector functions
  • Shows immune activation and significant antitumor activity
  • Potential for improved management of immune-related adverse events (irAEs)
  • For research use only
Catalog No. 50-004-23141