Learn More
Strem, An Ascensus Company CAS# 93166-53-1. 1g. (N,N-Diethyldithiocarbamato)dimethylgold(III), 97% (99.999%-Au) PURATREM.
SDP
Supplier:  Strem, An Ascensus Company 7917001G
Encompass
CAS# 93166-53-1. 1g. (N,N-Diethyldithiocarbamato)dimethylgold(III), 97% (99.999%-Au) PURATREM. . Molecular Weight: 375.30. Molecular Formula: (CH3)2Au(S2CN(C2H5)2). Color/form: yellow xtl. Strem# 79-1700. http://www.strem.com/catalog/v/79-1700/
Catalog No. 50-204-5414