Learn More
Enzo Life Sciences Auranofin, (100mg), CAS Number: 34031-32-8
SDP
Supplier:  Enzo Life Sciences BMLEI2060100
Encompass_Preferred
Anti-arthritic agent. Alternative Name: SKF 39162, 1-Thio-beta-D-glucopyranosatotriethyl phosphine gold-2,3,4,6-tetraacetate. Formula: C20H34AuO9PS. MW: 678.5. CAS Number: 34031-32-8. MI: 14: 878. Purity: ≥98% (HPLC). Appearance: White solid. Solubility: Soluble in DMSO (5mg/ml) or 100% ethanol (4mg/ml). Use/Stability: Stable for at least 1 year after receipt when stored, as supplied, at room temperature. Stock solutions are stable for up to 3 months at -20°C. Long Term Storage: Ambient
Catalog No. NC0693485