Learn More
Medchemexpress LLC HY-B1123 50mg , Auranofin CAS:34031-32-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB112350MG
Encompass_Preferred
HY-B1123 50mg Auranofin CAS: 34031-32-8 Auranofin (SKF-39162) is a thioredoxin reductase (TrxR) inhibitor with an IC50 of 0.2 μM. Auranofin exhibits antiviral activity against SARS-CoV21, with a CC50 of 4.2 μM for monkey kidney Vero E6 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9402