Learn More
Medchemexpress LLC HY-N2388 10mg Medchemexpress, Auraptene CAS:495-02-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN238810MG
Encompass_Preferred
Medchemexpress, HY-N2388 10mg Auraptene CAS:495-02-3 Auraptene is the most abundant naturally occurring geranyloxycoumarin. Auraptene is primarily isolated from plants in the Rutaceae family, such as citrus fruits. Auraptene decreases the secretion of matrix metalloproteinase 2 ( MMP-2 ) as well as key inflammatory mediators, including IL-6, IL-8, and chemokine (C-C motif) ligand-5(CCL5) [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3926